288012
Dimethyltin dichloride
97%
Synonym(s):
Dichlorodimethylstannane, Dichlorodimethyltin
About This Item
Recommended Products
Assay
97%
form
solid
mp
101-106 °C (lit.)
SMILES string
C[Sn](C)(Cl)Cl
InChI
1S/2CH3.2ClH.Sn/h2*1H3;2*1H;/q;;;;+2/p-2
InChI key
PKKGKUDPKRTKLJ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1
Target Organs
Immune system,Nervous system
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service