Merck
A low-density DNA microarray for analysis of markers in breast cancer.
The International journal of biological markers (2002-04-09)
M Lacroix, N Zammatteo, J Remacle, G Leclercq
RESUMEN
Breast cancer remains a major cause of death in women from Western countries. In the near future, advances in both nucleic acids technology and tumor biology should be widely exploited to improve the diagnosis, prognosis, and outcome prediction of this disease. The DNA microarray, also called biochip, is a promising tool for performing massive, simultaneous, fast, and standardized analyses of multiple molecular markers in tumor samples. However, most currently available microarrays are expensive, which is mainly due to the amount (several thousands) of different DNA capture sequences that they carry. While these high-density microarrays are best suited for basic studies, their introduction into the clinical routine remains hypothetical. We describe here the principles of a low-density microarray, carrying only a few hundreds of capture sequences specific to markers whose importance in breast cancer is generally recognized or suggested by the current medical literature. We provide a list of about 250 of these markers. We also examine some potential difficulties (homologies between marker and/or variant sequences, size of sequences, etc.) associated with the production of such a low-cost microarray.