Merck
Effect of p120 catenin silencing on biological behaviors of PANC-1 cells.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban (2012-10-18)
Zhangjun Cheng, Volker Assfag, Xin Shi, Shibo Lin, Jiangyan Xia, Pinghua Yang, Norbert Hüser, Feng Shen
RESUMEN
This study examined the possible role of p120ctn in the pathogenesis and development of pancreatic cancer. PANC-1 cells, a kind of human pancreatic carcinoma cell line, were cultured in this study. p120ctn was immunocytochemically detected in PANC-1 cells. The recombinant lentivirus vector was constructed to knock down the p120ctn expression of PANC-1 cells. Real-time quantitative PCR (RQ-PCR) and Western blotting were used to determine the expression of p120ctn and E-cadherin in PANC-1 cells after p120ctn knockdown. The adhesion, invasion and migration capacity of PANC-1 cells after p120ctn knockdown was detected by cell adhesion, invasion and migration assays. Cell growth was measured by the MTT method. Cell cycle and apoptosis were analyzed by fluorescence-activated cell sorting. The results showed that p120ctn knockdown led to significantly down-regulated E-cadherin and a reduced cell-to-cell adhesion ability in PANC-1 cells. shRNA-mediated knockdown of p120ctn reduced invasion and migration capacity of PANC-1 cells, inhibited cell growth, caused a significant decrease in the percentage of cells in G(1), an increase in S, and promoted apoptosis of PANC-1 cells. It was concluded that p120ctn plays a pivotal role in the proliferation and metastasis of pancreatic carcinoma, suggesting that p120ctn is a novel target for pancreatic carcinoma treatment.