Merck
  • Molecular mechanism of metal-independent decomposition of organic hydroperoxides by halogenated quinoid carcinogens and the potential biological implications.
Molecular mechanism of metal-independent decomposition of organic hydroperoxides by halogenated quinoid carcinogens and the potential biological implications.
Chemical research in toxicology (2015-03-20)
Chun-Hua Huang, Fu-Rong Ren, Guo-Qiang Shan, Hao Qin, Li Mao, Ben-Zhan Zhu
RESUMEN
Halogenated quinones (XQ) are a class of carcinogenic intermediates and newly identified chlorination disinfection byproducts in drinking water. Organic hydroperoxides (ROOH) can be produced both by free radical reactions and enzymatic oxidation of polyunsaturated fatty acids. ROOH have been shown to decompose to alkoxyl radicals via catalysis by transition metal ions, which may initiate lipid peroxidation or transform further to the reactive aldehydes. However, it is not clear whether XQ react with ROOH in a similar manner to generate alkoxyl radicals metal-independently. By complementary applications of ESR spin-trapping, HPLC/high resolution mass spectrometric and other analytical methods, we found that 2,5-dichloro-1,4-benzoquinone (DCBQ) could significantly enhance the decomposition of a model ROOH tert-butylhydroperoxide, resulting in the formation of t-butoxyl radicals independent of transition metals. On the basis of the above findings, we detected and identified, for the first time, an unprecedented C-centered quinone ketoxy radical. Then, we extended our study to the more physiologically relevant endogenous ROOH 13-hydroperoxy-9,11-octadecadienoic acid and found that DCBQ could also markedly enhance its decomposition to generate the reactive lipid alkyl radicals and the genotoxic 4-hydroxy-2-nonenal (HNE). Similar results were observed with other XQ. In summary, these findings demonstrated that XQ can facilitate ROOH decomposition to produce reactive alkoxyl, quinone ketoxy, lipid alkyl radicals, and genotoxic HNE via a novel metal-independent mechanism, which may explain partly their potential genotoxicity and carcinogenicity.