Merck
Adsorption and activation of molecular oxygen over atomic copper(I/II) site on ceria.
Nature communications (2020-08-13)
Liqun Kang, Bolun Wang, Qiming Bing, Michal Zalibera, Robert Büchel, Ruoyu Xu, Qiming Wang, Yiyun Liu, Diego Gianolio, Chiu C Tang, Emma K Gibson, Mohsen Danaie, Christopher Allen, Ke Wu, Sushila Marlow, Ling-Dong Sun, Qian He, Shaoliang Guan, Anton Savitsky, Juan J Velasco-Vélez, June Callison, Christopher W M Kay, Sotiris E Pratsinis, Wolfgang Lubitz, Jing-Yao Liu, Feng Ryan Wang
RESUMEN
Supported atomic metal sites have discrete molecular orbitals. Precise control over the energies of these sites is key to achieving novel reaction pathways with superior selectivity. Here, we achieve selective oxygen (O2) activation by utilising a framework of cerium (Ce) cations to reduce the energy of 3d orbitals of isolated copper (Cu) sites. Operando X-ray absorption spectroscopy, electron paramagnetic resonance and density-functional theory simulations are used to demonstrate that a [Cu(I)O2]3- site selectively adsorbs molecular O2, forming a rarely reported electrophilic η2-O2 species at 298 K. Assisted by neighbouring Ce(III) cations, η2-O2 is finally reduced to two O2-, that create two Cu-O-Ce oxo-bridges at 453 K. The isolated Cu(I)/(II) sites are ten times more active in CO oxidation than CuO clusters, showing a turnover frequency of 0.028 ± 0.003 s-1 at 373 K and 0.01 bar PCO. The unique electronic structure of [Cu(I)O2]3- site suggests its potential in selective oxidation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Copper(II) 2-ethylhexanoate
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad