Merck
ASS234, As a New Multi-Target Directed Propargylamine for Alzheimer's Disease Therapy.
Frontiers in neuroscience (2016-07-23)
José Marco-Contelles, Mercedes Unzeta, Irene Bolea, Gerard Esteban, Rona R Ramsay, Alejandro Romero, Ricard Martínez-Murillo, M Carmo Carreiras, Lhassane Ismaili
RESUMEN
ASS2324 is a hybrid compound resulting from the juxtaposition of donepezil and the propargylamine PF9601N ASS2324 is a multi-target directed propargylamine able to bind to all the AChE/BuChE and MAO A/B enzymesASS2324 shows antioxidant, neuroprotective and suitable permeability propertiesASS2324 restores the scopolamine-induced cognitive impairment to the same extent as donepezil, and is less toxicASS2324 prevents β-amyloid induced aggregation in the cortex of double transgenic miceASS2324 is the most advanced anti-Alzheimer agent for pre-clinical studies that we have identified in our laboratories The complex nature of Alzheimer's disease (AD) has prompted the design of Multi-Target-Directed Ligands (MTDL) able to bind to diverse biochemical targets involved in the progress and development of the disease. In this context, we have designed a number of MTD propargylamines (MTDP) showing antioxidant, anti-beta-amyloid, anti-inflammatory, as well as cholinesterase and monoamine oxidase (MAO) inhibition capacities. Here, we describe these properties in the MTDL ASS234, our lead-compound ready to enter in pre-clinical studies for AD, as a new multipotent, permeable cholinesterase/monoamine oxidase inhibitor, able to inhibit Aβ-aggregation, and possessing antioxidant and neuroprotective properties.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
ASS234, ≥98% (HPLC)