Merck
Impact of the fermentation parameters pH and temperature on stress resilience of Lactobacillus reuteri DSM 17938.
AMB Express (2019-05-19)
Armando Hernández, Christer U Larsson, Radoslaw Sawicki, Ed W J van Niel, Stefan Roos, Sebastian Håkansson
RESUMEN
This study was undertaken to investigate the impact of culture pH (4.5-6.5) and temperature (32-37 °C) on the stress resilience of Lactobacillus reuteri DSM 17938 during freeze-drying and post freeze-drying exposure to low pH (pH 2) and bile salts. Response-surface methodology analysis revealed that freeze-drying survival rates [Formula: see text] were linearly related to pH with the highest survival rate of 80% when cells were cultured at pH 6.5 and the lowest was 40% when cells were cultured at pH 4.5. The analysis further revealed that within the chosen temperature range the culture temperature did not significantly affect the freeze-drying survival rate. However, fermentation at pH 4.5 led to better survival rates when rehydrated cells were exposed to low pH shock or bile salts. Thus, the effect of pH on freeze-drying survival was in contrast to effects on low pH and bile salts stress tolerance. The rationale behind this irreconcilability is based on the responses being dissimilar and are not tuned to each other. Culturing strain DSM 17938 at pH values higher than 5.5 could be a useful option to improve the survivability and increase viable cell numbers in the final freeze-dried product. However, the dissimilar responses for the process- and application parameters tested here suggest that an optimal compromise has to be found in order to obtain the most functional probiotic product possible.