Merck
Natural occurrence of genotoxic and carcinogenic alkenylbenzenes in Indonesian jamu and evaluation of consumer risks.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2018-05-05)
Suparmi Suparmi, Diana Widiastuti, Sebastiaan Wesseling, Ivonne M C M Rietjens
RESUMEN
The consumer risks of jamu, Indonesian traditional herbal medicines, was assessed focussing on the presence of alkenylbenzene containing botanical ingredients. Twenty-three out of 25 samples contained alkenylbenzenes at levels ranging from 3.8 to 440 μg/kg, with methyleugenol being the most frequently encountered alkenylbenzene. The estimated daily intake (EDI) resulting from jamu consumption was estimated to amount to 0.2-171 μg/kg bw/day for individual alkenylbenzenes, to 0.9-203 μg/kg bw/day when adding up all alkenylbenzenes detected, and to 0.9-551 μg/kg bw/day when expressed in methyleugenol equivalents using interim relative potency (REP) factors. The margin of exposure (MOE) values obtained were generally <10,000 indicating a priority for risk management when assuming daily consumption during a lifetime. Using Haber's rule it was estimated that two weeks consumption of these jamu only once would not raise a concern (MOE >10,000). However, when considering use for two weeks every year during a lifetime, 5 samples still raise a concern. It is concluded that the consumption of alkenylbenzene containing jamu can be of concern especially when consumed on a daily basis for longer periods of time on a regular basis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Eugenol methyl ether, 99%