Key Documents
Z763985
StripSpin 12 mini centrifuge, round rotor
round rotor, 8 position
About This Item
UNSPSC Code:
41103909
NACRES:
NB.22
feature
to hold 8 x 1.5/2.0 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific C1008-ROT
parameter
5500 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models StripSpin 12 mini
General description
Retro fit 8-position rotor to hold 1.5/2.0ml tubes in Benchmark C1008 mini centrifuge