Key Documents
Z690481
Ace replacement reactor body
capacity 500 mL, PTFE
About This Item
UNSPSC Code:
41103817
NACRES:
NB.51
material
PTFE
manufacturer/tradename
Ace Glass 6391-05
capacity
500 mL
Categorías relacionadas
General description
Ace reactor bodies are replacement parts for the Ace all-PTFE filter reactor assembly, Z690368, Z690473 and Z690503.