Key Documents
Z125326
Ace PTFE ferrules
for tubing o.d., 6.4 mm (1/4 in.)
About This Item
UNSPSC Code:
27121703
NACRES:
NB.35
feature
for tubing o.d. 6.4 mm (1/4 in.)
manufacturer/tradename
Ace Glass 1171007
General description
For use in place of O-rings on threaded glassware.