HPLC Analysis of Oxazepam Enantiomers on Astec® CYCLOBOND I 2000 DNP
![HPLC Analysis of Oxazepam Enantiomers on Astec® CYCLOBOND I 2000 DNP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/301/774/4b242a61-c21d-4c16-b85a-45da32b2c730/800/4b242a61-c21d-4c16-b85a-45da32b2c730.jpg)
CONDITIONS
column
CYCLOBOND I 2000 DNP, 25 cm x 4.6 mm I.D., 5 μm particles (25024AST)
mobile phase
[A] acetonitrile; [B] 20 mM ammonium phosphate, pH 2.9; (20:80, A:B)
flow rate
1 mL/min
column temp.
5 °C
detector
UV, 254 nm
injection
3 μL
sample
oxazepam, 1 mg/mL in acetonitrile:water (50:50)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null