HPLC Analysis of Monoclonal Antibodies (mAb) with Charge Variance on Antibodix® WCX-NP10
CONDITIONS
column
Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 μm particles (Z777272)
mobile phase
(A) 10 mM sodium phosphate buffer, pH 7.5; (B) A + 100 mM NaCl
gradient
15 to 55% B in 30 min
flow rate
0.8 mL/min
pressure
840 psi (58 bar)
column temp.
ambient
detector
UV, 214 nm
injection
5 μL
sample
monoclonal antibody
Description
Legal Information
Antibodix is a registered trademark of Sepax Technologies