HPLC Analysis of Bupivacaine Enantiomers on Astec® CHIROBIOTIC® V2, Acetate Mobile Phase
![HPLC Analysis of Bupivacaine Enantiomers on Astec® CHIROBIOTIC® V2, Acetate Mobile Phase application for HPLC](/deepweb/assets/sigmaaldrich/product/images/381/155/44834b56-73b3-4c03-9a05-b941ff52d6b0/800/44834b56-73b3-4c03-9a05-b941ff52d6b0.jpg)
Materials
analytical column
Product No.
Description
Pricing
Application
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
[A] methanol; [B] 20 mM ammonium acetate, pH 4.1; (80:20, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 205 nm
injection
10 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC