Merck
  • The selective reaction of aryl halides with KOH: synthesis of phenols, aromatic ethers, and benzofurans.
The selective reaction of aryl halides with KOH: synthesis of phenols, aromatic ethers, and benzofurans.
Journal of the American Chemical Society (2006-08-17)
Kevin W Anderson, Takashi Ikawa, Rachel E Tundel, Stephen L Buchwald
ABSTRACT
The direct and selective synthesis of phenols from aryl/heteroaryl halides and KOH has been achieved through the use of highly active monophosphine-based catalysts derived from Pd(2)dba(3) and ligands L1 or L2 and the biphasic solvent system 1,4-dioxane/H(2)O. We have also demonstrated a one-pot method of phenol formation/alkylation for the preparation of alkyl aryl ethers from aryl halides. In many instances, this protocol overcomes limitations in existing Pd-catalyzed coupling reactions of aliphatic alcohols with aryl halides. Finally, we demonstrate that substituted benzofurans can be prepared efficiently via a Pd-catalyzed phenol formation/cyclization protocol starting from 2-chloroaryl alkynes.
Sigma-Aldrich
Me4tButylXphos, 96%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
tBuXPhos, 98%