Merck
  • Connexin 43 enhances paclitaxel cytotoxicity in colorectal cancer cell lines.
Connexin 43 enhances paclitaxel cytotoxicity in colorectal cancer cell lines.
Experimental and therapeutic medicine (2017-08-16)
Siqi Wang, Shiwu Zhang, Zhenying Zhao, Chunze Zhang, Xiaoyun Yang, Yijia Wang
ABSTRACT
Colorectal cancer has a relatively low sensitivity to paclitaxel. The purpose of this study was to investigate the role of connexin 43 (Cx43), which is a structural component of gap junctional communication (GJC), in paclitaxel cytotoxicity in colorectal cancer cells. Three colorectal cancer cell lines (HCT106, HCT116 and LoVo) were transfected with Cx43 and used to examine paclitaxel cytotoxicity. A western blot assay was used to confirm Cx43 expression in transfected cell lines as well as the expression of several proteins that are associated with paclitaxel cytotoxicity. A parachute dye-coupling assay was used to measure GJC function. An MTT assay was used to analyze the viability of paclitaxel-treated cells. Cx43 expression level and GJC function were significantly upregulated by the transfection (P<0.05). The viability of transfected cells was significantly inhibited compared with that of untransfected cells when treated with paclitaxel (20 or 80 nM) at high culture density but not at low culture density (P<0.05). Cx43 transfection significantly increased the mitotic arrest, tubulin polymerization and apoptosis effects of paclitaxel (P<0.05). It was also found that paclitaxel had an inhibitory effect on GJC function after 12 h of treatment in LoVo cells (P<0.05). These results indicate that Cx43 may serve as a target of paclitaxel chemotherapy for colorectal cancer.
Sigma-Aldrich
Anti-Tubulin α antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Connexin-43 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (Fc specific), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Survivin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Cyclin B1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Caspase 3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution