- Graphene quantum dots-terbium ions as novel sensitive and selective time-resolved luminescent probes.
Graphene quantum dots-terbium ions as novel sensitive and selective time-resolved luminescent probes.
We propose an alternative approach for the development of analytical methods based on terbium-sensitized luminescence (TSL). TSL is based on the complexation between Tb(III) ions and fluorescent organic compounds that have appropriate functional groups to complex with Tb(III). We report the use of graphene quantum dot (GQDs) nanoparticles to improve the sensitivity and selectivity of TSL detection. GQDs can react with terbium ions through the carboxylic groups present in their structure. These Tb(III)-GQD complexes, formed in situ in aqueous solution, can be used as time-resolved luminescent probes. Ascorbic acid was selected as a target analyte to demonstrate the suitability of the proposed method. The selectivity of the TSL method was highly improved for most of the interferences tested. Under the optimum conditions [Tb(III) concentration 5 × 10