Merck
  • Direct Measurement of Single-Molecule Adenosine Triphosphatase Hydrolysis Dynamics.
Direct Measurement of Single-Molecule Adenosine Triphosphatase Hydrolysis Dynamics.
ACS nano (2017-12-08)
Jie Li, Gen He, Ueno Hiroshi, Wenzhe Liu, Hiroyuki Noji, Chuanmin Qi, Xuefeng Guo
ABSTRACT
F
Sigma-Aldrich
Disilane, electronic grade
Sigma-Aldrich
Diborane, 99.99% (diborane only), 9-11% (balance hydrogen), 10% in hydrogen, electronic grade
Sigma-Aldrich
p-Phenylene diisothiocyanate, 98%