Merck
  • Self-calibrated fluorescent probe resembled as an indicator of the lysosomal phosphatase pertaining to the cancer cells.
Self-calibrated fluorescent probe resembled as an indicator of the lysosomal phosphatase pertaining to the cancer cells.
Journal of photochemistry and photobiology. B, Biology (2017-11-08)
Arup Podder, Susan M Alex, Mrinmoy Maiti, Kaustabh Kumar Maiti, Sankarprasad Bhuniya
ABSTRACT
A self-calibrated fluorescent probe Lyso-Phos has synthesized followed by a straightforward synthetic pathway. Lyso-Phos acts as an indicator for lysosomal phosphatase. Its photophysical property including cellular imaging was described. Lyso-Phos showed ratiometric UV-Vis- absorption changes from λ
Sigma-Aldrich
Diethyl chlorophosphate, 97%
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%