Merck
  • The effect of liquid hot water pretreatment on the chemical-structural alteration and the reduced recalcitrance in poplar.
The effect of liquid hot water pretreatment on the chemical-structural alteration and the reduced recalcitrance in poplar.
Biotechnology for biofuels (2017-12-08)
Mi Li, Shilin Cao, Xianzhi Meng, Michael Studer, Charles E Wyman, Arthur J Ragauskas, Yunqiao Pu
ABSTRACT
Hydrothermal pretreatment using liquid hot water (LHW) is capable of substantially reducing the cell wall recalcitrance of lignocellulosic biomass. It enhances the saccharification of polysaccharides, particularly cellulose, into glucose with relatively low capital required. Due to the close association with biomass recalcitrance, the structural change of the components of lignocellulosic materials during the pretreatment is crucial to understand pretreatment chemistry and advance the bio-economy. Although the LHW pretreatment has been extensively applied and studied, the molecular structural alteration during pretreatment and its significance to reduced recalcitrance have not been well understood. We investigated the effects of LHW pretreatment with different severity factors (log This study revealed that the solubilization of xylan, the reduction of hemicellulose molecular weights and cellulose degree of polymerization, and the cleavage of alkyl-aryl ether bonds in lignin resulted from LHW pretreatment are critical factors associated with reduced cell wall recalcitrance. The chemical-structural changes of the three major components, cellulose, lignin, and hemicellulose, during LHW pretreatment provide useful and fundamental information of factors governing feedstock recalcitrance during hydrothermal pretreatment.
Sigma-Aldrich
Allyl methyl sulfone, 96%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Peracetic acid solution, 32 wt. % in dilute acetic acid