Merck
  • Development and validation of a method for the determination of regulated fragrance allergens by High-Performance Liquid Chromatography and Parallel Factor Analysis 2.
Development and validation of a method for the determination of regulated fragrance allergens by High-Performance Liquid Chromatography and Parallel Factor Analysis 2.
Journal of chromatography. A (2017-10-24)
Jessica Pérez-Outeiral, Saioa Elcoroaristizabal, Jose Manuel Amigo, Maider Vidal
ABSTRACT
This work presents the development and validation of a multivariate method for quantitation of 6 potentially allergenic substances (PAS) related to fragrances by ultrasound-assisted emulsification microextraction coupled with HPLC-DAD and PARAFAC2 in the presence of other 18 PAS. The objective is the extension of a previously proposed univariate method to be able to determine the 24 PAS currently considered as allergens. The suitability of the multivariate approach for the qualitative and quantitative analysis of the analytes is discussed through datasets of increasing complexity, comprising the assessment and validation of the method performance. PARAFAC2 showed to adequately model the data facing up different instrumental and chemical issues, such as co-elution profiles, overlapping spectra, unknown interfering compounds, retention time shifts and baseline drifts. Satisfactory quality parameters of the model performance were obtained (R
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Hydroxycitronellal, ≥95%, FCC, FG
Sigma-Aldrich
Benzyl cinnamate, 99%
Sigma-Aldrich
2-Dodecanol, 99%