Merck
  • Premature termination codon readthrough in human cells occurs in novel cytoplasmic foci and requires UPF proteins.
Premature termination codon readthrough in human cells occurs in novel cytoplasmic foci and requires UPF proteins.
Journal of cell science (2017-07-27)
Jieshuang Jia, Elisabeth Werkmeister, Sara Gonzalez-Hilarion, Catherine Leroy, Dieter C Gruenert, Frank Lafont, David Tulasne, Fabrice Lejeune
ABSTRACT
Nonsense-mutation-containing messenger ribonucleoprotein particles (mRNPs) transit through cytoplasmic foci called P-bodies before undergoing nonsense-mediated mRNA decay (NMD), a cytoplasmic mRNA surveillance mechanism. This study shows that the cytoskeleton modulates transport of nonsense-mutation-containing mRNPs to and from P-bodies. Impairing the integrity of cytoskeleton causes inhibition of NMD. The cytoskeleton thus plays a crucial role in NMD. Interestingly, disruption of actin filaments results in both inhibition of NMD and activation of premature termination codon (PTC) readthrough, while disruption of microtubules causes only NMD inhibition. Activation of PTC readthrough occurs concomitantly with the appearance of cytoplasmic foci containing UPF proteins and mRNAs with nonsense mutations but lacking the P-body marker DCP1a. These findings demonstrate that in human cells, PTC readthrough occurs in novel 'readthrough bodies' and requires the presence of UPF proteins.
Sigma-Aldrich
MISSION® esiRNA, targeting human UPF2
Sigma-Aldrich
MISSION® esiRNA, targeting human UPF3B
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-GFP, N-terminal antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution