Merck
  • Metformin represses cancer cells via alternate pathways in N-cadherin expressing vs. N-cadherin deficient cells.
Metformin represses cancer cells via alternate pathways in N-cadherin expressing vs. N-cadherin deficient cells.
Oncotarget (2015-09-12)
Rongbin Ge, Zongwei Wang, Shulin Wu, Yangjia Zhuo, Aleksandar G Otsetov, Chao Cai, Weide Zhong, Chin-Lee Wu, Aria F Olumi
ABSTRACT
Metformin has emerged as a potential anticancer agent. Here, we demonstrate that metformin plays an anti-tumor role via repressing N-cadherin, independent of AMPK, in wild-type N-cadherin cancer cells. Ectopic-expression of N-cadherin develops metformin-resistant cancer cells, while suppression of N-cadherin sensitizes cancer to metformin. Manipulation of AMPK expression does not alter sensitivity of cancer to metformin. We show that NF-kappaB is a downstream molecule of N-cadherin and metformin regulates NF-kappaB signaling via suppressing N-cadherin. Moreover, we also suggest that TWIST1 is an upstream molecule of N-cadherin/NF-kappaB signaling and manipulation of TWIST1 expression changes the sensitivity of cancer cells to metformin. In contrast to the cells that express N-cadherin, in N-cadherin deficient cells, metformin plays an anti-tumor role via activation of AMPK. Ectopic expression of N-cadherin makes cancer more resistant to metformin. Therefore, we suggest that metformin's anti-cancer therapeutic effect is mediated through different molecular mechanism in wild-type vs. deficient N-cadherin cancer cells. At last, we selected 49 out of 984 patients' samples with prostatic cancer after radical prostatectomy (selection criteria: Gleason score ≥ 7 and all patients taking metformin) and showed levels of N-cadherin, p65 and AMPK could predict post-surgical recurrence in prostate cancer after treatment of metformin.
Sigma-Aldrich
Bay 11-7085, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-N-Cadherin antibody, Mouse monoclonal, clone GC-4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-FLIP Antibody, CT, Upstate®, from rabbit
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder