Merck
  • The Baylis-Hillman acetates as a valuable source for one-pot multistep synthesis: a facile synthesis of functionalized tri-/tetracyclic frameworks containing azocine moiety.
The Baylis-Hillman acetates as a valuable source for one-pot multistep synthesis: a facile synthesis of functionalized tri-/tetracyclic frameworks containing azocine moiety.
Organic letters (2007-06-01)
Deevi Basavaiah, Kunche Aravindu
ABSTRACT
The Baylis-Hillman acetates have been conveniently transformed into tri-/tetracyclic heterocyclic frameworks containing an important azocine moiety via one-pot multistep protocol involving alkylation, reduction, and cyclization sequence.