Merck
  • N-Cadherin Regulates Cell Migration Through a Rab5-Dependent Temporal Control of Macropinocytosis.
N-Cadherin Regulates Cell Migration Through a Rab5-Dependent Temporal Control of Macropinocytosis.
Traffic (Copenhagen, Denmark) (2016-04-12)
Meng-Hsuan Wen, Jen-Yeu Wang, Yu-Ting Chiu, Mei-Pin Wang, Sue-Ping Lee, Chin-Yin Tai
ABSTRACT
Macropinocytosis is a clathrin-independent endocytic pathway implicated in fluid uptake, pathogen invasion and cell migration. During collective cell migration, macropinocytosis occurs primarily at membrane ruffles arising from the leading edges of migrating cells. We report here that N-cadherin (Ncad) regulates the tempo of macropinocytosis and thereby influences wound-induced collective cell migration. Using live-cell and super-resolution imaging techniques, we observed that Ncad formed clusters at the membrane ruffles and macropinosomes. De-clustering of Ncad by an interfering antibody impaired the recruitment of Rab5-an early endosomal marker-to the macropinosomes. Moreover, we demonstrated that Ncad interacts with Rab5, and laser ablation of Ncad caused Rab5 to dissociate from the macropinosomes. Although Rab5 detached from macropinosomes upon the de-clustering of Ncad, the recruitment of late endosomal marker Rab7 occurred earlier. Consequently, both centripetal trafficking of macropinosomes and collective migration were accelerated due to de-clustering of Ncad. Thus, our results suggest that Ncad is involved in the maturation of macropinocytosis through Rab5 recruitment, linking macropinocytosis and cell migration through a novel function of Ncad.
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Anti-N-Cadherin antibody, Mouse monoclonal, clone GC-4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-RAB5C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution