Merck
  • Altered neuronal markers following treatment with mood stabilizer and antipsychotic drugs indicate an increased likelihood of neurotransmitter release.
Altered neuronal markers following treatment with mood stabilizer and antipsychotic drugs indicate an increased likelihood of neurotransmitter release.
Clinical psychopharmacology and neuroscience : the official scientific journal of the Korean College of Neuropsychopharmacology (2013-02-23)
Elizabeth Scarr, Brian Dean
ABSTRACT
Given the ability of mood stabilizers and antipsychotics to promote cell proliferation, we wanted to determine the effects of these drugs on neuronal markers previously reported to be altered in subjects with psychiatric disorders. Male Sprauge-Dawley rats were treated with vehicle (ethanol), lithium (25.5 mg per day), haloperidol (0.1 mg/kg), olanzapine (1.0 mg/kg) or a combination of lithium and either of the antipsychotic drugs for 28 days. Levels of cortical synaptic (synaptosomal associated protein-25, synaptophysin, vesicle associated protein and syntaxin) and structural (neural cell adhesion molecule and alpha-synuclein) proteins were determined in each treatment group using Western blots. Compared to the vehicle treated group; animals treated with haloperidol had greater levels of synaptosomal associated protein-25 (p<0.01) and neural cell adhesion molecule (p<0.05), those treated with olanzapine had greater levels of synaptophysin (p<0.01) and syntaxin (p<0.01). Treatment with lithium alone did not affect the levels of any of the proteins. Combining lithium and haloperidol resulted in greater levels of synaptophysin (p<0.01), synaptosomal associated protein-25 (p<0.01) and neural cell adhesion molecule (p<0.01). The combination of lithium and olanzapine produced greater levels of synaptophysin (p<0.01) and alpha-synuclein (p<0.05). Lithium alone had no effect on the neuronal markers. However, haloperidol and olanzapine affected different presynaptic markers. Combining lithium with olanzapine additionally increased alpha-synuclein. These drug effects need to be taken into account by future studies examining presynaptic and neuronal markers in tissue from subjects with psychiatric disorders.
Sigma-Aldrich
Monoclonal Anti-Neural Cell Adhesion Molecule antibody produced in mouse, clone NCAM-0B11, ascites fluid
Sigma-Aldrich
Anti-Synaptobrevin Antibody, clone SP10, ascites fluid, clone SP10, Chemicon®
Sigma-Aldrich
Anti-Syntaxin Antibody, clone SP8, ascites fluid, clone SP8, Chemicon®