Merck
Downregulation of urea transporter UT-A1 activity by 14-3-3 protein.
American journal of physiology. Renal physiology (2015-05-23)
Xiuyan Feng, Zenggang Li, Yuhong Du, Haian Fu, Janet D Klein, Hui Cai, Jeff M Sands, Guangping Chen
ABSTRACT
Urea transporter (UT)-A1 in the kidney inner medulla plays a critical role in the urinary concentrating mechanism and thereby in the regulation of water balance. The 14-3-3 proteins are a family of seven isoforms. They are multifunctional regulatory proteins that mainly bind to phosphorylated serine/threonine residues in target proteins. In the present study, we found that all seven 14-3-3 isoforms were detected in the kidney inner medulla. However, only the 14-3-3 γ-isoform was specifically and highly associated with UT-A1, as demonstrated by a glutathione-S-transferase-14-3-3 pulldown assay. The cAMP/adenylyl cyclase stimulator forskolin significantly enhanced their binding. Coinjection of 14-3-3γ cRNA into oocytes resulted in a decrease of UT-A1 function. In addition, 14-3-3γ increased UT-A1 ubiquitination and protein degradation. 14-3-3γ can interact with both UT-A1 and mouse double minute 2, the E3 ubiquitin ligase for UT-A1. Thus, activation of cAMP/PKA increases 14-3-3γ interactions with UT-A1 and stimulates mouse double minute 2-mediated UT-A1 ubiquitination and degradation, thereby forming a novel regulatory mechanism of urea transport activity.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-14-3-3σ Antibody, clone CS112-2A8, clone CS112-2A8, Upstate®, from mouse
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Rabbit Anti-Chicken IgY Antibody, HRP conjugate, Upstate®, from rabbit