Merck
  • Expedited palladium-catalyzed amination of aryl nonaflates through the use of microwave-irradiation and soluble organic amine bases.
Expedited palladium-catalyzed amination of aryl nonaflates through the use of microwave-irradiation and soluble organic amine bases.
The Journal of organic chemistry (2006-01-04)
Rachel E Tundel, Kevin W Anderson, Stephen L Buchwald
ABSTRACT
[reaction: see text] Microwave-assisted, palladium-catalyzed C-N bond-forming reactions with aryl/heteroaryl nonaflates and amines using the soluble amine bases DBU (1,8-diazabicyclo[5.4.0]undec-7-ene) or MTBD (7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene) and ligands (1-3) resulted in good to excellent yields (71-99%) of arylamines in short reaction times (1-45 min).
Sigma-Aldrich
7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene, 98%