Merck
  • Infrared microspectroscopy detects protein misfolding cyclic amplification (PMCA)-induced conformational alterations in hamster scrapie progeny seeds.
Infrared microspectroscopy detects protein misfolding cyclic amplification (PMCA)-induced conformational alterations in hamster scrapie progeny seeds.
The Journal of biological chemistry (2013-10-29)
Martin L Daus, Katja Wagenführ, Achim Thomzig, Susann Boerner, Peter Hermann, Antje Hermelink, Michael Beekes, Peter Lasch
ABSTRACT
The self-replicative conformation of misfolded prion proteins (PrP) is considered a major determinant for the seeding activity, infectiousness, and strain characteristics of prions in different host species. Prion-associated seeding activity, which converts cellular prion protein (PrP(C)) into Proteinase K-resistant, infectious PrP particles (PrP(TSE)), can be monitored in vitro by protein misfolding cyclic amplification (PMCA). Thus, PMCA has been established as a valuable analytical tool in prion research. Currently, however, it is under discussion whether prion strain characteristics are preserved during PMCA when parent seeds are amplified in PrP(C) substrate from the identical host species. Here, we report on the comparative structural analysis of parent and progeny (PMCA-derived) PrP seeds by an improved approach of sensitive infrared microspectroscopy. Infrared microspectroscopy revealed that PMCA of native hamster 263K scrapie seeds in hamster PrP(C) substrate caused conformational alterations in progeny seeds that were accompanied by an altered resistance to Proteinase K, higher sedimentation velocities in gradient ultracentrifugations, and a longer incubation time in animal bioassays. When these progeny seeds were propagated in hamsters, misfolded PrP from brain extracts of these animals showed mixed spectroscopic and biochemical properties from both parental and progeny seeds. Thus, strain modifications of 263K prions induced by PMCA seem to have been partially reversed when PMCA products were reinoculated into the original host species.
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, for molecular biology, ≥800 units/mL
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥500 units/mL, buffered aqueous glycerol solution
Sigma-Aldrich
Proteinase K from Tritirachium album, ≥3.0 unit/mg solid, lyophilized powder
Sigma-Aldrich
Proteinase K from Tritirachium album, Reagents designed and manufactured under current ISO 13485 certification.