Merck
  • Dried blood spots as a sample collection technique for the determination of pharmacokinetics in clinical studies: considerations for the validation of a quantitative bioanalytical method.
Dried blood spots as a sample collection technique for the determination of pharmacokinetics in clinical studies: considerations for the validation of a quantitative bioanalytical method.
Analytical chemistry (2009-01-22)
Neil Spooner, Rakesh Lad, Matt Barfield
ABSTRACT
A novel approach has been developed for the quantitative determination of circulating drug concentrations in clinical studies using dried blood spots (DBS) on paper, rather than conventional plasma samples. A quantitative bioanalytical HPLC-MS/MS assay requiring small blood volumes (15 microL) has been validated using acetaminophen as a tool compound (range 25 to 5000 ng/mL human blood). The assay employed simple solvent extraction of a punch taken from the DBS sample, followed by reversed phase HPLC separation, combined with selected reaction monitoring mass spectrometric detection. In addition to performing routine experiments to establish the validity of the assay to internationally accepted criteria (precision, accuracy, linearity, sensitivity, selectivity), a number of experiments were performed to specifically demonstrate the quality of the quantitative data generated using this novel sample format, namely, stability of the analyte and metabolites in whole human blood and in DBS samples; effect of the volume of blood spotted, the device used to spot the blood, or the temperature of blood spotted. The validated DBS approach was successfully applied to a clinical study (single oral dose of 500 mg or 1 g acetaminophen).
Product Description
QIAcard FTA DMPK Cards, FTA DMPK-C card, 4 sample areas per card, pack of 100 cards
QIAcard FTA DMPK Cards, FTA DMPK-B card, 4 sample areas per card, pack of 100 cards
QIAcard FTA DMPK Cards, FTA DMPK-A card, 4 sample areas per card, pack of 100 cards