Merck
  • Lipopolysaccharide and aldoheptose biosynthesis in transketolase mutants of Salmonella typhimurium.
Lipopolysaccharide and aldoheptose biosynthesis in transketolase mutants of Salmonella typhimurium.
Proceedings of the National Academy of Sciences of the United States of America (1971-08-01)
L Eidels, M J Osborn
ABSTRACT
Genetic and biochemical evidence that sedoheptulose-7-phosphate is an obligatory precursor of the L-glycero-D-mannoheptose residues of the lipopolysaccharide of Salmonella was obtained by isolation and characterization of transketolase-negative mutants of Salmonella typhimurium. These mutants, which are defective in synthesis of sedoheptulose-7-phosphate, were found to produce an incomplete heptose-deficient lipopolysaccharide, and were also sensitive to bile salts, a characteristic property of heptose-deficient mutants. Phenotypic repair of the defect in lipopolysaccharide synthesis was obtained by addition of exogenous sedoheptulose-7-phosphate to growing cultures of the mutant strains. Characterization of revertants isolated either as transketolase-positive or heptose-positive provided further evidence that the heptose deficiency resulted from mutation at the transketolase locus. On the basis of these findings a possible pathway for conversion of sedoheptulose-7-phosphate to L-glycero-D-mannoheptose is proposed.
Sigma-Aldrich
D-Sedoheptulose 7-phosphate lithium salt, ā‰„90% (TLC)