Merck
  • Delay of natural bone loss by higher intakes of specific minerals and vitamins.
Delay of natural bone loss by higher intakes of specific minerals and vitamins.
Critical reviews in food science and nutrition (2001-06-13)
A Schaafsma, P J de Vries, W H Saris
ABSTRACT
For early prevention or inhibition of postmenopausal and age-related bone loss, nutritional interventions might be a first choice. For some vitamins and minerals an important role in bone metabolism is known or suggested. Calcium and vitamin D support bone mineral density and are basic components in most preventive strategies. Magnesium is involved in a number of activities supporting bone strength, preservation, and remodeling. Fluorine and strontium have bone-forming effects. However, high amounts of both elements may reduce bone strength. Boron is especially effective in case of vitamin D, magnesium, and potassium deficiency. Vitamin K is essential for the activation of osteocalcin. Vitamin C is an important stimulus for osteoblast-derived proteins. Increasing the recommended amounts (US RDA 1989), adequate intakes (US DRI 1997), or assumed normal intakes of mentioned food components may lead to a considerable reduction or even prevention of bone loss, especially in late postmenopausal women and the elderly.
Sigma-Aldrich
Strontium, random pieces, 99%
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Strontium, granular, 99% trace metals basis