- Rational design of a dual chemosensor for cyanide anion sensing based on dicyanovinyl-substituted benzofurazan.
Rational design of a dual chemosensor for cyanide anion sensing based on dicyanovinyl-substituted benzofurazan.
The Journal of organic chemistry (2011-11-18)
Zhipeng Liu, Xiaoqing Wang, Zhenghao Yang, Weijiang He
PMID22088170
ABSTRACT
A dicyanovinyl-substituted benzofurazan derivative (C1) was prepared as an efficient ratiometric chemosensor for cyanide anion detection in aqueous acetonitrile solution. Mechanism studies suggested that the nucleophilic addition of cyanide to the Ī±-position of the dicyanovinyl group blocked the ICT progress of C1 and induced remarkable emission and absorption shift.
MATERIALS