Merck
  • Fabrication of single-phase titanium carbide layers from MWCNTs using high DC pulse.
Fabrication of single-phase titanium carbide layers from MWCNTs using high DC pulse.
Nanotechnology (2010-01-07)
Woo Sik Kim, Sook Young Moon, Jeong Hoon Lee, Sin Young Bang, Bong Geun Choi, Heon Ham, Tohru Sekino, Kwang Bo Shim
ABSTRACT
Single-phase layered titanium carbide (TiC) was successfully synthesized by reacting carbon nanotubes (CNTs) and titanium dioxide (TiO2) under a high direct current (DC) pulse. Single-phase TiC layer fabrication is confirmed as the transformation of multi-layered graphene from MWCNTs. Therefore its thickness and width is almost identical to those of transformed graphene layers. This is the first report on the formation of single-phase layered nano-TiC. Scanning electron microscopy (SEM), x-ray diffraction (XRD), x-ray photoelectron spectroscopy (XPS) and high-resolution transmission electron microscopy (HR-TEM) were used for the characteristic analysis of single-phase layered TiC structures.
Sigma-Aldrich
Titanium(IV) carbide, powder, <4 μm, ≥99% (Ti)
Sigma-Aldrich
Titanium(IV) carbide, −325 mesh, 98%
Sigma-Aldrich
Titanium carbide, nanopowder, ≤200 nm particle size (SEM)