Merck
  • Generalizing the breakdown of the maximum hardness and minimum polarizabilities principles for nontotally symmetric vibrations to non-pi-conjugated organic molecules.
Generalizing the breakdown of the maximum hardness and minimum polarizabilities principles for nontotally symmetric vibrations to non-pi-conjugated organic molecules.
The journal of physical chemistry. A (2006-07-13)
Miquel Torrent-Sucarrat, Miquel Duran, Josep M Luis, Miquel Solà
ABSTRACT
In previous works we have shown that certain pi-conjugated organic molecules possess nontotally symmetric vibrations that break the maximum hardness (MHP) and minimum polarizability principles (MPP). We have also derived a set of simple rules to determine a priori without calculations whether a particular pi-conjugated organic molecule violates these two principles. In the present work, we generalize these results, and we show that not only pi-conjugated organic molecules but also other molecules without pi-conjugated structure or even pi-bonds can exhibit nontotally symmetric molecular distortions that do not follow these two principles. We have also found that the breakdowns of the MHP and the MPP are not necessarily connected, since the polarizability is not always proportional to the softness. Finally, we also introduced a methodology based on the diagonalization of the hardness Hessian matrix with respect to the vibrational normal coordinates to determine the nontotally symmetric molecular displacements that do not follow the MHP.
Sigma-Aldrich
Diborane, 99.99% (diborane only), 9-11% (balance hydrogen), 10% in hydrogen, electronic grade