Merck
  • Polymer-based lanthanide luminescent sensor for detection of the hydrolysis product of the nerve agent Soman in water.
Polymer-based lanthanide luminescent sensor for detection of the hydrolysis product of the nerve agent Soman in water.
Analytical chemistry (1999-02-09)
A L Jenkins, O M Uy, G M Murray
ABSTRACT
The techniques of molecular imprinting and sensitized lanthanide luminescence have been combined to create the basis for a sensor that can selectively measure the hydrolysis product of the nerve agent Soman in water. The sensor functions by selectively and reversibly binding the phosphonate hydrolysis product of this agent to a functionality-imprinted copolymer possessing a coordinatively bound luminescent lanthanide ion, Eu3+. Instrumental support for this device is designed to monitor the appearance of a narrow luminescence band in the 610-nm region of the Eu3+ spectrum that results when the analyte is coordinated to the copolymer. The ligand field shifted luminescence was excited using 1 mW of the 465.8-nm line of an argon ion laser and monitored via an optical fiber using a miniature spectrometer. For this configuration, the limit of detection for the hydrolysis product is 7 parts per trillion (ppt) in solution with a linear range from 10 ppt to 10 ppm. Chemical and spectroscopic selectivities have been combined to reduce the likelihood of false positive analyses. Chemically analogous organophosphorus pesticides tested against the sensor have been shown to not interfere with determination.
Sigma-Aldrich
Methyl 3,5-dimethylbenzoate, 98%