Merck
  • High-throughput flow cytometry compatible biosensor based on fluorogen activating protein technology.
High-throughput flow cytometry compatible biosensor based on fluorogen activating protein technology.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2013-01-11)
Yang Wu, Phillip H Tapia, Gregory W Fisher, Alan S Waggoner, Jonathan Jarvik, Larry A Sklar
ABSTRACT
Monitoring the trafficking of multiple proteins simultaneously in live cells is of great interest because many receptor proteins are found to function together with others in the same cell. However, existing fluorescent labeling techniques have restricted the mechanistic study of functional receptor pairs. We have expanded a hybrid system combining fluorogen-activating protein (FAP) technology and high-throughput flow cytometry to a new type of biosensor that is robust, sensitive, and versatile. This provides the opportunity to study multiple trafficking proteins in the same cell. Human beta2 adrenergic receptor (β2AR) fused with FAP AM2.2 and murine C-C chemokines receptor type 5 fused with FAP MG13 was chosen for our model system. The function of the receptor and the binding between MG13 and fluorogen MG-2p have been characterized by flow cytometry and confocal microscopy assays. The binding of fluorogen and the FAP pair is highly specific, while both FAP-tagged fusion proteins function similarly to their wild-type counterparts. The system has successfully served as a counter screen assay to eliminate false positive compounds identified in a screen against NIH Molecular Libraries Small Molecule Repository targeting regulators of the human β2AR.
Sigma-Aldrich
Malachite Green solution, for microscopy
Sigma-Aldrich
Malachite Green oxalate salt, certified by the Biological Stain Commission, certified by the BSC
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution A, for microscopy
Sigma-Aldrich
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard