Merck
  • Phenomenon of quantum entanglement in a system composed of two minimal protocells.
Phenomenon of quantum entanglement in a system composed of two minimal protocells.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life (2012-12-18)
Arvydas Tamulis, Mantas Grigalavicius, Jonas Baltrusaitis
ABSTRACT
The quantum mechanical self-assembly of two separate photoactive supramolecular systems with different photosynthetic centers was investigated by means of density functional theory methods. Quantum entangled energy transitions from one subsystem to the other and the assembly of logically controlled artificial minimal protocells were modeled. The systems studied were based on different photoactive sensitizer molecules covalently bonded to a non-canonical oxo-guanine::cytosine supramolecule with the precursor of a fatty acid (pFA) molecule attached via Van der Waals forces, all surrounded by water molecules. The electron correlation interactions responsible for the weak hydrogen and Van der Waals chemical bonds increased due to the addition of polar water solvent molecules. The distances between the separated sensitizer, nucleotide, pFA, and water molecules are comparable to Van der Waals and hydrogen bonding radii. As a result, the overall system becomes compressed, resulting in photo-excited electron tunneling from the sensitizer (bis(4-diphenylamine-2-phenyl)-squarine or 1,4-bis(N,N-dimethylamino)naphthalene) to the pFA molecules. Absorption spectra as well as electron transfer trajectories associated with the different excited states were calculated using time dependent density functional theory methods. The results allow separation of the quantum entangled photosynthetic transitions within the same minimal protocell and with the neighboring minimal protocell. The transferred electron is used to cleave a "waste" organic molecule resulting in the formation of the desired product. A two variable, quantum entangled AND logic gate was proposed, consisting of two input photoactive sensitizer molecules and one output (pFA molecule). It is proposed that a similar process might be applied for the destruction of tumor cancer cells or to yield building blocks in artificial cells.
Product Description
Supelco
1-Naphthylamine, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
1-Naphthylamine, 97%
Pricing and availability is not currently available.
Millipore
Nitrate Reagent A, suitable for microbiology
Pricing and availability is not currently available.