Merck
  • Pentaquinone based probe for nanomolar detection of zinc ions: chemosensing ensemble as an antioxidant.
Pentaquinone based probe for nanomolar detection of zinc ions: chemosensing ensemble as an antioxidant.
Dalton transactions (Cambridge, England : 2003) (2012-11-21)
Vandana Bhalla, Roopa, Manoj Kumar
ABSTRACT
A pentaquinone based compound 3a has been synthesized which undergoes significant fluorescence enhancement in the presence of Zn(2+) ions with a detection limit up to nanomolar range in THF. Further, the zinc ensemble of 3a is evaluated for its anti-oxidizing property which is better than commercially available antioxidants.
Product Description
Supelco
1,4-Benzoquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
p-Benzoquinone, for spectrophotometric det. of amines, ≥99.5% (HPLC)
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%