Merck
  • [The clinical application of perfluorotributylamine liquid in the complicated retinal detachment].
[The clinical application of perfluorotributylamine liquid in the complicated retinal detachment].
Hunan yi ke da xue xue bao = Hunan yike daxue xuebao = Bulletin of Hunan Medical University (2002-05-23)
X Zhu, D Jiang
ABSTRACT
To evaluate the clinical application value of perfluorotributylamine liquid (PFTL) made in China in the complicated retinal detachment. PFTL was injected into eyeball after vitreoretinal surgery, it could open funnel of retinal detachment, push subretinal fluid out from peripheral hole and flat and immobilize the posterior retinal during vitreoretinal surgery. In eyes with giant tears, PFTL could unfold and fix retinal flap. The retinas in all cases with giant tears were reattached, and sixty per cent in the eyes with PVR-D2-D3 reattached. In 8 eyes the visual acuity was improved. PFTL is valuable in treatment of complicated retinal detachment. It facilitates vitreoretinal surgery, obviates the potential complications.
Product Description
Supelco
Perfluorotributylamine (PFTBA), analytical standard
Supelco
Heptacosafluorotributylamine, analytical standard, suitable for mass spectrometry (MS)
Sigma-Aldrich
Heptacosafluorotributylamine, liquid