Merck
  • A high-throughput-compatible, fluorescence anisotropy-based assay for ATP-dependent supercoiled DNA relaxation by human topoisomerase IIα.
A high-throughput-compatible, fluorescence anisotropy-based assay for ATP-dependent supercoiled DNA relaxation by human topoisomerase IIα.
Biochemical pharmacology (2013-02-19)
Adam B Shapiro
ABSTRACT
A novel, high-throughput-compatible assay for the ATP-dependent supercoiled DNA relaxing activity of human topoisomerase IIα (hTopoIIα) is described. The principle of detection is the preferential binding of the oligodeoxyribonucleotide BODIPY-TMR-5'-TTCTTCTTCT-3' to relaxed double-stranded plasmid containing the triplex forming sequence (TTC)9 versus the supercoiled plasmid. Binding of the oligonucleotide to the plasmid increases the fluorescence anisotropy of the BODIPY-TMR label. Optimization of the assay conditions was conducted to maximize the signal and the activity of the topoisomerase. The multiwell assay plate-based fluorescence anisotropy assay gave the same values for the potencies of several previously reported inhibitors of hTopoIIα as a gel electrophoresis-based assay of DNA relaxation.
Sigma-Aldrich
DNA Gyrase from Escherichia coli, aqueous glycerol solution
Sigma-Aldrich
Topoisomerase II α human, (Single band on SDS-PAGE), liquid