Merck
  • Contactin-associated protein-like 2 (CNTNAP2) mutations impair the essential α-secretase cleavages, leading to autism-like phenotypes.
Contactin-associated protein-like 2 (CNTNAP2) mutations impair the essential α-secretase cleavages, leading to autism-like phenotypes.
Signal transduction and targeted therapy (2024-03-01)
Qing Zhang, Mengen Xing, Zhengkai Bao, Lu Xu, Yang Bai, Wanqi Chen, Wenhao Pan, Fang Cai, Qunxian Wang, Shipeng Guo, Jing Zhang, Zhe Wang, Yili Wu, Yun Zhang, Jia-Da Li, Weihong Song
ABSTRACT
Mutations in the Contactin-associated protein-like 2 (CNTNAP2) gene are associated with autism spectrum disorder (ASD), and ectodomain shedding of the CNTNAP2 protein plays a role in its function. However, key enzymes involved in the C-terminal cleavage of CNTNAP2 remain largely unknown, and the effect of ASD-associated mutations on this process and its role in ASD pathogenesis remain elusive. In this report we showed that CNTNAP2 undergoes sequential cleavages by furin, ADAM10/17-dependent α-secretase and presenilin-dependent γ-secretase. We identified that the cleavage sites of ADAM10 and ADAM17 in CNTNAP2 locate at its C-terminal residue I79 and L96, and the main α-cleavage product C79 by ADAM10 is required for the subsequent γ-secretase cleavage to generate CNTNAP2 intracellular domain (CICD). ASD-associated CNTNAP2 mutations impair the α-cleavage to generate C79, and the inhibition leads to ASD-like repetitive and social behavior abnormalities in the Cntnap2-I1254T knock-in mice. Finally, exogenous expression of C79 improves autism-like phenotypes in the Cntnap2-I1254T knock-in and Cntnap2-/- knockout mice. This data demonstrates that the α-secretase is essential for CNTNAP2 processing and its function. Our study indicates that inhibition of the cleavage by pathogenic mutations underlies ASD pathogenesis, and upregulation of its C-terminal fragments could have therapeutical potentials for ASD treatment.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CNTNAP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution