Merck
  • Andrographolide Relieves Post-Operative Wound Pain but Affects Local Angiogenesis.
Andrographolide Relieves Post-Operative Wound Pain but Affects Local Angiogenesis.
Pharmaceuticals (Basel, Switzerland) (2022-12-24)
Yi-Lo Lin, Jiunn-Wang Liao, Shunching Wang, Badrinathan Sridharan, Hsin-Ju Lee, Ai Li, Kai-Ming Chang, Ching-Yang Wu, Siendong Huang, Kai-Ting Chang, Dinesh Chandra Agrawal, Ching-Jung Chen, Meng-Jen Lee
ABSTRACT
Andrographolide (Andro), the major constituent of Andrographis paniculata Nees (Acanthaceae), is was known to reduces inflammatory reaction. In the current study, the ability of Andro to reduce pain sensation in a rat post-operative wound model was explored. The hind paws of 18 Sprague-Dawley rats (SD) bearing post-operative wounds received the following three treatments: Saline, Andro via direct injection into the paw (Andro-injected) and Tablet containing Andro + poly (lactic-co-glycolic acid) (PLGA) (Andro-tablet). Von Frey tests assessed mechanical allodynia at 1, 3, 5 h and 1-, 2-, 3-, 4-, and 5-days post-operation. Behavioral analyses were performed to measure reaction threshold and reaction frequencies. Immunoreactivity of p-ERK and GluR1 was examined in the dorsal horn of the spinal cord. Histopathological and immunostaining studies were conducted on paw epidermis to observe the gross morphology and angiogenesis. The threshold for inducing allodynia increased and the reaction frequency reduced in the Andro-injected group compared to the saline-group, at 3 h post-surgery and the effect lasted between 3-4 days. The threshold for inducing pain and reaction frequency for the Andro-tablet group did not differ from the saline-treated group. The levels of p-ERK and GluR1 in the dorsal horn were reduced after Andro treatment. No significant difference in wound healing index was observed between saline and Andro-injected groups, but CD-31 staining showed less angiogenesis in the Andro-injected group. Andro significantly reduced mechanical allodynia compared to saline treatment, both in shorter and longer time frames. Furthermore, Andro influenced the expression of p-ERK and GluR1 in the dorsal horn, and the angiogenesis process in the wound healing area.
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution