Merck
  • Loss of CRY2 promotes regenerative myogenesis by enhancing PAX7 expression and satellite cell proliferation.
Loss of CRY2 promotes regenerative myogenesis by enhancing PAX7 expression and satellite cell proliferation.
MedComm (2023-01-14)
Yingxue Hao, Ting Xue, Song-Bai Liu, Sha Geng, Xinghong Shi, Panting Qian, Wei He, Jiqing Zheng, Yanfang Li, Jing Lou, Tianze Shi, Ge Wang, Xiaoxiao Wang, Yanli Wang, Yangxin Li, Yao-Hua Song
ABSTRACT
The regenerative capacity of skeletal muscle is dependent on satellite cells. The circadian clock regulates the maintenance and function of satellite cells. Cryptochrome 2 (CRY2) is a critical component of the circadian clock, and its role in skeletal muscle regeneration remains controversial. Using the skeletal muscle lineage and satellite cell-specific CRY2 knockout mice (CRY2scko), we show that the deletion of CRY2 enhances muscle regeneration. Single myofiber analysis revealed that deletion of CRY2 stimulates the proliferation of myoblasts. The differentiation potential of myoblasts was enhanced by the loss of CRY2 evidenced by increased expression of myosin heavy chain (MyHC) and myotube formation in CRY2-/- cells versus CRY2+/+ cells. Immunostaining revealed that the number of mononucleated paired box protein 7 (PAX7+) cells associated with myotubes formed by CRY2-/- cells was increased compared with CRY2+/+ cells, suggesting that more reserve cells were produced in the absence of CRY2. Loss of CRY2 leads to the activation of the ERK1/2 signaling pathway and ETS1, which binds to the promoter of PAX7 to induce its transcription. CRY2 deficient myoblasts survived better in ischemic muscle. Therefore, CRY2 is essential in regulating skeletal muscle repair.
Sigma-Aldrich
Anti-phospho-ETS1 (pThr38) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Cry2 (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution