Merck
  • Effects of Extraction Methods and Conditions on Bioactive Compounds Extracted from Phaeodactylum tricornutum.
Effects of Extraction Methods and Conditions on Bioactive Compounds Extracted from Phaeodactylum tricornutum.
Acta chimica Slovenica (2021-02-04)
Saniye AkyÄ L, IĹźÄ L Ä Lter, Mehmet KoĂ, Zeliha Demirel, AyĹźegĂĽl ErdoÄźan, Meltem Conk Dalay, Figen Kaymak Ertekin
ABSTRACT
The effect of homogenization, ultrasound and microwave extraction methods and conditions on fucoxanthin content, total phenolic content and antioxidant activity of extracts obtained from Phaeodactylum tricornutum were investigated in this study. The solvent/biomass ratio was the most effective parameter on fucoxanthin content, total phenolic content and antioxidant activity. The maximum fucoxanthin content (5.60 ± 0.06 mg/g) and antioxidant activity (763.00 ± 15.88 EC50 ?g/mL extract) were obtained with the homogenization extraction method whose optimum conditions were 1.93% biomass/solvent ratio, ~5200 rpm homogenization rate and 14.2 min extraction time. Although the ultrasonic extraction method has reached the approximately same level of fucoxanthin content (5.24 ± 0.07 mg/g)), TPC (67.68 ± 1.58 mg gallic acid/L) and antioxidant activity (619.90 ± 17.16 EC50 ?g/mL extract) at an amplitude of 55.72%, a higher biomass/ solvent ratio (2.72%) and a longer extraction time (17.37 min) have been required. The lowest fucoxanthin content, total phenolic content and antioxidant activity were determined for the microwave extraction method.
Sigma-Aldrich
Neoxanthin, ≥90% (HPLC)