Merck
  • Poly(ADP-ribose) polymerase 1 inhibition protects against low shear stress induced inflammation.
Poly(ADP-ribose) polymerase 1 inhibition protects against low shear stress induced inflammation.
Biochimica et biophysica acta (2012-10-23)
Wei-dong Qin, Shu-jian Wei, Xu-ping Wang, Juan Wang, Wen-ke Wang, Fuqiang Liu, Lei Gong, Fei Yan, Yun Zhang, Mingxiang Zhang
ABSTRACT
Atherosclerosis begins as local inflammation of vessels at sites of disturbed flow, where low shear stress (LSS) leads to mechanical irritation and plaque development and progression. Nuclear enzyme poly(ADP-ribose) polymerase 1 (PARP-1) is associated with the inflammation response during atherosclerosis. We investigated the role and underlying mechanism of PARP-1 in LSS-induced inflammation in human umbilical vein endothelial cells (HUVECs). HUVECs were simulated by LSS (0.4Pa). PARP-1 expression was inhibited by ABT888 or siRNA. The inducible nitric oxide synthase (iNOS) and intercellular adhesion molecular-1 (ICAM-1) expression was regulated by LSS in a time dependent manner. LSS could increase superoxide production and 3-nitrotyrosine formation. LSS induced DNA damage as assessed by H2A.X phosphorylation and comet assay. Compared with cells under static, LSS increased PARP-1 expression and PAR formation via MEK/ERK signaling pathway. PARP-1 inhibition increased Sirt1 activity through an increased intracellular nicotinamide adenine dinucleotide (NAD(+)) level. Moreover, PARP-1 inhibition attenuated LSS-induced iNOS and ICAM-1 upregulation by inhibiting nuclear factor kappa B (NF-κB) nuclear translocation and activity, with a reduced NF-κB phosphorylation. LSS induced oxidative damage and PARP-1 activation via MEK/ERK pathway. PARP-1 inhibition restored Sirt1 activity by increasing NAD(+) level and decreased iNOS and ICAM-1 expression by inhibiting NF-κB nuclear translocation and activity as well as NF-κB phosphorylation. PARP-1 played a fundamental role in LSS induced inflammation. Inhibition of PARP-1 might be a mechanism for treatment of inflammation response during atherosclerosis.
Sigma-Aldrich
PARP-1 human, recombinant, expressed in E. coli
Pricing and availability is not currently available. Contact Technical Service