Merck
  • Targeted magnetic nanoparticle hyperthermia for the treatment of oral cancer.
Targeted magnetic nanoparticle hyperthermia for the treatment of oral cancer.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology (2019-07-17)
Christopher J Legge, Helen E Colley, Michelle A Lawson, Andrea E Rawlings
ABSTRACT
Patients with oral squamous cell carcinoma currently experience a five-year survival rate of approximately 60% with conventional surgical, chemotherapy and radiotherapy treatments. Magnetic hyperthermia offers an alternative treatment method by utilising the heating properties of magnetic nanoparticles to produce thermal ablation of the tumour site when exposed to an alternating magnetic field. In this study, we investigate in vitro if targeted magnetic hyperthermia offers a potential treatment for oral squamous cell carcinoma. Magnetic iron oxide nanoparticles, with a biocompatible silica coating, were produced and conjugated with antibodies to target integrin αvβ6, a well-characterised oral squamous cell carcinoma biomarker. Utilising the heating properties of the magnetic nanoparticles, we exposed them to an alternating magnetic field to produce thermo ablation of tumour cells either negative for or overexpressing integrin αvβ6. The cell surface biomarker, αvβ6 integrin, was upregulated in tissue biopsies from oral squamous cell carcinoma patients compared to normal tissue. Functionalisation of the silica coating with anti-αvβ6 antibodies enabled direct targeting of the nanoparticles to αvβ6 overexpressing cells and applying thermal therapy significantly increased killing of the targeted tumour cells compared to control cells. Combining antibody-targeting magnetic nanoparticles with thermal ablation offers a promising therapy for the targeted treatment of oral squamous cell carcinoma.
Sigma-Aldrich
Anti-Integrin αVβ6 Antibody, clone E7P6, azide free, clone E7P6, Chemicon®, from mouse