Merck
  • Dorsal raphe serotonergic neurons promote arousal from isoflurane anesthesia.
Dorsal raphe serotonergic neurons promote arousal from isoflurane anesthesia.
CNS neuroscience & therapeutics (2021-05-12)
Ao Li, Rui Li, Pengrong Ouyang, Huihui Li, Sa Wang, Xinxin Zhang, Dan Wang, Mingzi Ran, Guangchao Zhao, Qianzi Yang, Zhenghua Zhu, Hailong Dong, Haopeng Zhang
ABSTRACT
General anesthesia has been widely applied in surgical or nonsurgical medical procedures, but the mechanism behind remains elusive. Because of shared neural circuits of sleep and anesthesia, whether serotonergic system, which is highly implicated in modulation of sleep and wakefulness, regulates general anesthesia as well is worth investigating. Immunostaining and fiber photometry were used to assess the neuronal activities. Electroencephalography spectra and burst-suppression ratio (BSR) were used to measure anesthetic depth and loss or recovery of righting reflex to indicate the induction or emergence time of general anesthesia. Regulation of serotonergic system was achieved through optogenetic, chemogenetic, or pharmacological methods. We found that both Fos expression and calcium activity were significantly decreased during general anesthesia. Activation of 5-HT neurons in the dorsal raphe nucleus (DRN) decreased the depth of anesthesia and facilitated the emergence from anesthesia, and inhibition deepened the anesthesia and prolonged the emergence time. Furthermore, agonism or antagonism of 5-HT 1A or 2C receptors mimicked the effect of manipulating DRN serotonergic neurons. Our results demonstrate that 5-HT neurons in the DRN play a regulative role of general anesthesia, and activation of serotonergic neurons could facilitate emergence from general anesthesia partly through 5-HT 1A and 2C receptors.
Sigma-Aldrich
M100907, ≥98% (HPLC)
Sigma-Aldrich
LP44, ≥98% (HPLC), solid
Sigma-Aldrich
WAY-100635 maleate salt, powder
Sigma-Aldrich
SB-269970 hydrochloride, powder, ≥98% (HPLC)
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
SB-399885 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
CP-93129 dihydrochloride hydrate, ≥98% (HPLC)