Merck
  • The therapeutic efficacy of water-soluble coenzyme Q10 in an experimental model of tacrolimus-induced diabetes mellitus.
The therapeutic efficacy of water-soluble coenzyme Q10 in an experimental model of tacrolimus-induced diabetes mellitus.
The Korean journal of internal medicine (2020-04-14)
Yi Quan, Kang Luo, Sheng Cui, Sun Woo Lim, Yoo Jin Shin, Eun Jeong Ko, Ju Hwan Kim, Sang J Chung, Soo Kyung Bae, Byung Ha Chung, Chul Woo Yang
ABSTRACT
Coenzyme Q10 (CoQ10) has antioxidant effects and is commercially available and marketed extensively. However, due to its low bioavailability, its effects are still controversial. We developed a water-soluble CoQ10-based micelle formulation (CoQ10-W) and tested it in an experimental model of tacrolimus (TAC)-induced diabetes mellitus (DM). We developed CoQ10-W from a glycyrrhizic-carnitine mixed layer CoQ10 micelle preparation based on acyltransferases. TAC-induced DM rats were treated with either lipid-soluble CoQ10 (CoQ10-L) or CoQ10-W for 4 weeks. Their plasma and pancreatic CoQ10 concentrations were measured using liquid chromatography- tandem mass spectrometry. The therapeutic efficacies of CoQ10-W and CoQ10-L on TAC-induced DM were compared using functional and morphological parameters and their effects on cell viability and reactive oxygen species (ROS) production were also evaluated in cultured rat insulinoma cells. The plasma CoQ10 level was significantly increased in the CoQ10-W group compared to that in the CoQ10-L group. Intraperitoneal glucose tolerance tests and glucose-stimulated insulin secretion revealed that CoQ10-W controlled hyperglycemia and restored insulin secretion significantly better than CoQ10-L. The TAC-mediated decrease in pancreatic islet size was significantly attenuated by CoQ10-W but not by CoQ10-L. TAC-induced oxidative stress and apoptosis were significantly more reduced by CoQ10-W than CoQ10-L. Electron microscopy revealed that CoQ10-W restored TAC-induced attenuation in the number of insulin granules and the average mitochondrial area, unlike CoQ10-L. In vitro studies showed that CoQ10-L and CoQ10-W both improved cell viability and reduced ROS production in TAC-treated islet cells to a similar extent. CoQ10-W has better therapeutic efficacy than CoQ10-L in TAC-induced DM.
Sigma-Aldrich
Trisodium glycyrrhizinate hydrate, AldrichCPR
Sigma-Aldrich
Monoclonal Anti-Insulin antibody produced in mouse, clone K36AC10, ascites fluid
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder