Merck
  • Shear-horizontal surface acoustic wave sensor for non-invasive monitoring of dynamic cell spreading and attachment in wound healing assays.
Shear-horizontal surface acoustic wave sensor for non-invasive monitoring of dynamic cell spreading and attachment in wound healing assays.
Biosensors & bioelectronics (2020-11-23)
Manuel S Brugger, Lukas G Schnitzler, Timo Nieberle, Achim Wixforth, Christoph Westerhausen
ABSTRACT
A Love-wave based biosensor is introduced for analyzing a standardized wound healing assay by observing cell growth and quantifying cell detachment processes. Utilizing the piezoelectric material LiTaO3 36° XY-cut with a thin SiO2-cover layer, shear horizontal surface acoustic waves (SAW) are excited and detected by a set of Interdigital Transducers. Epithelial cells, being cultivated on the substrate and invading the sensors delay line cause a phase shift in the transmitted SAW signal. This phase shift correlates exactly with the surface coverage of the invading cells. After wound healing, emerging fluctuations in the phase shift signal provide information about the cell growth in a confluent cell layer. Additionally, the signal slope allows to quantify the cell detachment process induced by apoptosis, necrosis or cell lysis substances, respectively. Furthermore, culture conditions like temperature or osmolality can be simultaneously monitored by SAW. Based on a theoretical approach and using FEM simulations, we identified the acoustoelectric interaction as the main reason for the phase shift in various frequency- and time-dependent studies. Our model is validated by experimental data and allows predicting the phase change caused by variations in the cell-substrate distance or the volume ratio of the nucleus and the complete cell.
Sigma-Aldrich
Terg-a-zyme® enzyme detergent, pack of 11 kg
MDCK-II Cell Line canine